Learn More
Biomatik Corporation Mouse Phenylalanine-4-hydroxylase (PAH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37565
Encompass_Preferred
Mouse Phenylalanine-4-Hydroxylase (Pah) Elisa Kit for detecting Phenylalanine Hydroxylase in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P16331; Pah. Target Synonyms: Pah; Phenylalanine-4-hydroxylase; PAH; EC 1.14.16.1; Phe-4-monooxygenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8626