Learn More
Biomatik Corporation Mouse Phosphotylinosital 3 kinase, PI3K ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37577
Encompass_Preferred
Mouse Phosphotylinosital 3 Kinase, Pi3K Elisa Kit for detecting Phosphoinositide-3-Kinase, Catalytic, Alpha Polypeptide in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P42337; Pik3ca. Target Synonyms: Pik3ca; Phosphatidylinositol 4; 5-bisphosphate 3-kinase catalytic subunit alpha isoform; PI3-kinase subunit alpha; PI3K-alpha; PI3Kalpha; PtdIns-3-kinase subunit alpha; EC 2.7.1.153; Phosphatidylinositol 4; 5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha; PtdIns-3-kinase subunit p110-alpha; p110alpha; Phosphoinositide-3-kinase catalytic alpha polypeptide; Serine/threonine protein kinase PIK3CA; EC 2.7.11.1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8629