Learn More
Biomatik Corporation Mouse plasma kallikrein (KLKB1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37584
Encompass_Preferred
Mouse Plasma Kallikrein (Klkb1) Elisa Kit for detecting Plasma Kallikrein(Klkb1) in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200ng/ml. Sensitivity: 0.78ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P26262; Klkb1. Target Synonyms: Klkb1; Klk3; PkPlasma kallikrein; EC 3.4.21.34; Fletcher factor; Kininogenin; Plasma prekallikrein) [Cleaved into: Plasma kallikrein heavy chain; Plasma kallikrein light chain]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8630