Learn More
Biomatik Corporation Mouse procollagen N-terminal peptide, PNP ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37616
Encompass_Preferred
Mouse Procollagen I N-Terminal Peptide, Pinp Elisa Kit for detecting Procollagen I N-Terminal Peptide, PInp in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 4.7-300pg/ml. Sensitivity: 1.17pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: procollagen 1 N-terminal peptide; P1NP; procollagen I N-terminal peptide; PINP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8637