Learn More
Biomatik Corporation Mouse prostacycline, PGI2 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37639
Encompass_Preferred
Mouse Prostacycline, Pgi2 Elisa Kit for detecting Prostacycline, Pgi2 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 25-1000pg/ml. Sensitivity: 25pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8644