Learn More
Biomatik Corporation Mouse Protein notum homolog (NOTUM) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37669
Encompass_Preferred
Mouse Protein Notum Homolog (Notum) Elisa Kit for detecting Notum Pectinacetylesterase Homolog (Drosophila) in serum, plasma, tissue homogenates, cell culture supernates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8R116; Notum. Target Synonyms: NotumPalmitoleoyl-protein carboxylesterase NOTUM; EC 3.1.1.98. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8654