Learn More
Biomatik Corporation Mouse Proto-oncogene Wnt-1 (WNT1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37710
Encompass_Preferred
Mouse Proto-Oncogene Wnt-1 (Wnt1) Elisa Kit for detecting Wingless-Type Mmtv Integration Site Family, Member 1 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 18.75-1200pg/ml. Sensitivity: 4.68pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04426; Wnt1. Target Synonyms: Wnt1; Int-1; Wnt-1Proto-oncogene Wnt-1; Proto-oncogene Int-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8662