Learn More
Biomatik Corporation Mouse R-spondin-1 (RSPO1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37753
Encompass_Preferred
Mouse R-Spondin-1 (Rspo1) Elisa Kit for detecting R-Spondin Homolog (Xenopus Laevis) in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9Z132; Rspo1. Target Synonyms: Rspo1R-spondin-1; Cysteine-rich and single thrombospondin domain-containing protein 3; Cristin-3; mCristin-3; Roof plate-specific spondin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8670