Learn More
Biomatik Corporation Mouse Secretoglobin family 3A member 2 (SCGB3A2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37774
Encompass_Preferred
Mouse Secretoglobin Family 3A Member 2 (Scgb3A2) Elisa Kit for detecting Secretoglobin, Family 3A, Member 2 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 9.4-600pg/ml. Sensitivity: 2.35pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q920H1; Scgb3a2. Target Synonyms: Scgb3a2; Pnsp1; Ugrp1; Secretoglobin family 3A member 2; Pneumo secretory protein 1; PnSP-1; Uteroglobin-related protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8671