Learn More
Biomatik Corporation Mouse serum amyloid A-3 protein (SAA3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37808
Encompass_Preferred
Mouse Serum Amyloid A-3 Protein (Saa3) Elisa Kit for detecting Serum Amyloid A-3 Protein (Saa3) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04918; Saa3. Target Synonyms: Saa3; Serum amyloid A-3 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8676