Learn More
Biomatik Corporation Mouse Somatostatin, SS ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37848
Encompass_Preferred
Mouse Somatostatin, Ss Elisa Kit for detecting Somatostatin in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 4.7-300pg/ml. Sensitivity: 1.1pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P60041; Sst. Target Synonyms: Sst; Smst; Somatostatin [Cleaved into: Antrin; Somatostatin-28; Somatostatin-14; Neuronostatin; NST)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8678