Learn More
Biomatik Corporation Mouse syndecan 4 (SDC4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37879
Encompass_Preferred
Mouse Syndecan 4 (Sdc4) Elisa Kit for detecting Syndecan 4 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O35988; Sdc4. Target Synonyms: Sdc4; Syndecan-4; SYND4; Ryudocan core protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8687