Learn More
Biomatik Corporation Mouse Syndecan-1 (SDC1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37880
Encompass_Preferred
Mouse Syndecan-1 (Sdc1) Elisa Kit for detecting Syndecan 1 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P18828; Sdc1. Target Synonyms: Sdc1; Synd-1; Synd1; Syndecan-1; SYND1; CD antigen CD138. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8688