Learn More
Biomatik Corporation Mouse tartrate-resistant acid phosphatase 5b (TRACP-5b) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37884
Encompass_Preferred
Mouse Tartrate-Resistant Acid Phosphatase 5B (Tracp-5B) Elisa Kit for detecting Acid Phosphatase 5, Tartrate Resistant in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.078-5mIU/ml. Sensitivity: 0.02mIU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q05117; Acp5. Target Synonyms: Acp5; T5ap; Trap; Tartrate-resistant acid phosphatase type 5; TR-AP; EC 3.1.3.2; Tartrate-resistant acid ATPase; TrATPase; Type 5 acid phosphatase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8689