Learn More
Biomatik Corporation Mouse Thromboxane B2, TXB2 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37918
Encompass_Preferred
Mouse Thromboxane B2, Txb2 Elisa Kit for detecting Thromboxane B2, Txb2 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.35-150pg/ml. Sensitivity: 0.58pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8695