Learn More
Biomatik Corporation Mouse Thyroglobulin, TG ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37923
Encompass_Preferred
Mouse Thyroglobulin, Tg Elisa Kit for detecting Thyroglobulin in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 10-160ng/ml. Sensitivity: 10ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: O08710; Tg. Target Synonyms: Tg; Tgn; Thyroglobulin; Tg. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8696