Learn More
Biomatik Corporation Mouse -Secretase ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38077
Encompass_Preferred
Mouse Γ-Secretase Elisa Kit for detecting Anterior Pharynx Defective 1 Homolog B (C. Elegans) in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8C7N7; Aph1b. Target Synonyms: Aph1b; Gamma-secretase subunit APH-1B; APH-1b; Aph-1beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8717