Learn More
Biomatik Corporation Pig adiponectin, ADP ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38083
Encompass_Preferred
Pig Adiponectin, Adp Elisa Kit for detecting Adiponectin, Adp in serum, plasma, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 1.875-30ug/ml. Sensitivity: 1.875ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q6PP07. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8719