Learn More
Biomatik Corporation Pig C-Peptide ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38153
Encompass_Preferred
Pig C-Peptide Elisa Kit for detecting C-Peptide in serum, plasma, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 1.25-80ng/ml. Sensitivity: 0.312ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8729