Learn More
Biomatik Corporation Pig glucagon like peptide 2 (GLP2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38194
Encompass_Preferred
Pig Glucagon Like Peptide 2 (Glp2) Elisa Kit for detecting Glucagon Like Peptide 2(Glp2) in serum, plasma, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 2.4-150pg/ml. Sensitivity: 0.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8738