Learn More
Biomatik Corporation Pig Interleukin 6, IL-6 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38246
Encompass_Preferred
Pig Interleukin 6, Il-6 Elisa Kit for detecting Interleukin 6 (Interferon, Beta 2) in serum, plasma, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 1.25-80pg/ml. Sensitivity: 0.31pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P26893; IL6. Target Synonyms: IL6; Interleukin-6; IL-6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8747