Learn More
Biomatik Corporation Pig luteinizing hormone (LH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38270
Encompass_Preferred
Pig Luteinizing Hormone (Lh) Elisa Kit for detecting Luteinizing Hormone (Lh) in serum, plasma, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 3-600mIU/ml. Sensitivity: 1.5mIU/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P01232; LHB. Target Synonyms: LHBLutropin subunit beta; Lutropin beta chain; Luteinizing hormone subunit beta; LH-B; LSH-B; LSH-beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8754