Learn More
Biomatik Corporation Pig Procalcitonin, PCT ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38314
Encompass_Preferred
Pig Procalcitonin, Pct Elisa Kit for detecting Procalcitonin, Pct in serum, plasma, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 39-2500pg/ml. Sensitivity: 9.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8765