Learn More
Biomatik Corporation Pig serum amyloid A, SAA ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38330
Encompass_Preferred
Pig Serum Amyloid A, Saa Elisa Kit for detecting Serum Amyloid A, Saa in serum, plasma. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 19.53-5000pg/ml. Sensitivity: 19.53pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P0DSO0; SAA2. Target Synonyms: SAA2; Serum amyloid A-2 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8768