Learn More
Biomatik Corporation Pig Thyroid Stimulating Hormone, TSH ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38348
Encompass_Preferred
Pig Thyroid Stimulating Hormone, Tsh Elisa Kit for detecting Thyroid Stimulating Hormone, Tsh in serum, plasma. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.6-24uIU/ml. Sensitivity: 0.3uIU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01224; TSHB. Target Synonyms: TSHB; Thyrotropin subunit beta; Thyroid-stimulating hormone subunit beta; TSH-B; TSH-beta; Thyrotropin beta chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8773