Learn More
Biomatik Corporation Rabbit cross linked C-telopeptide of type collagen (CTX-) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38426
Encompass_Preferred
Rabbit Cross Linked C-Telopeptide Of Type Ii Collagen (Ctx-Ii) Elisa Kit for detecting Cross Linked C-Telopeptide Of Type II Collagen in serum, plasma, tissue homogenates. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: cross linked C-telopeptide of type 2 collagen (CTX-2); cross linked C-telopeptide of type II collagen (CTX-II). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8784