Learn More
Biomatik Corporation Rabbit plasmin-antiplasmin complex, PAP ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38516
Encompass_Preferred
Rabbit Plasmin-Antiplasmin Complex, Pap Elisa Kit for detecting Plasmin-Antiplasmin Complex, Pap in serum, plasma, tissue homogenates. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.25-16ng/ml. Sensitivity: 0.06ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8789