Learn More
Biomatik Corporation Rabbit Tartrate-resistant acid phosphatase 5b (TRAP5b) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38546
Encompass_Preferred
Rabbit Tartrate-Resistant Acid Phosphatase 5B (Trap5B) Elisa Kit for detecting Tartrate-Resistant Acid Phosphatase 5B (Trap5B) in serum, plasma, tissue homogenates. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 78-5000uU/ml. Sensitivity: 19.5uU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O97860; ACP5. Target Synonyms: ACP5; Tartrate-resistant acid phosphatase type 5; TR-AP; EC 3.1.3.2; Tartrate-resistant acid ATPase; TrATPase; Type 5 acid phosphatase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8793