Learn More
Biomatik Corporation Rabbit Vascular Endothelial cell Growth Factor, VEGF ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38562
Encompass_Preferred
Rabbit Vascular Endothelial Cell Growth Factor, Vegf Elisa Kit for detecting Vascular Endothelial Cell Growth Factor, Vegf in serum, plasma, tissue homogenates. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 1.56-100pg/ml. Sensitivity: 0.39pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8797