Learn More
Biomatik Corporation Rat 3-oxo-5-alpha-steroid 4-dehydrogenase 2 (SRD5A2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38579
Encompass_Preferred
Rat 3-Oxo-5-Alpha-Steroid 4-Dehydrogenase 2 (Srd5A2) Elisa Kit for detecting Steroid-5-Alpha-Reductase, Alpha Polypeptide 2 (3-Oxo-5 Alpha-Steroid Delta 4-Dehydrogenase Alpha 2) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6-1200pg/ml. Sensitivity: 3pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P31214; Srd5a2. Target Synonyms: Srd5a2; 3-oxo-5-alpha-steroid 4-dehydrogenase 2; 5 alpha-SR2; SR type 2; Steroid 5-alpha-reductase 2; S5AR 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8800