Learn More
Biomatik Corporation Rat Acylated Ghrelin (AG) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38608
Encompass_Preferred
Rat Acylated Ghrelin (Ag) Elisa Kit for detecting Acylated Ghrelin (Ag) in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8805