Learn More
Biomatik Corporation Rat advanced glycation end products, AGEs ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38623
Encompass_Preferred
Rat Advanced Glycation End Products, Ages Elisa Kit for detecting Advanced Glycation End Products, Ages in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.9-250ug/ml. Sensitivity: 0.97ug/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8806