Learn More
Biomatik Corporation Rat Alpha-fetoprotein, AFP ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38641
Encompass_Preferred
Rat Alpha-Fetoprotein, Afp Elisa Kit for detecting Alpha-Fetoprotein in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P02773; Afp. Target Synonyms: AfpAlpha-fetoprotein; Alpha-1-fetoprotein; Alpha-fetoglobulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8809