Learn More
Biomatik Corporation Rat Anti-Cyclic Citrullinated Peptide Antibody (Anti-CCP-antibody) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38678
Encompass_Preferred
Rat Anti-Cyclic Citrullinated Peptide Antibody (Anti-Ccp-Antibody) Elisa Kit for detecting Anti-Cyclic Citrullinated Peptide Antibody(Anti-Ccp-Antibody) in serum, plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12- 200IU/ml. Sensitivity: 0.78IU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8817