Learn More
Biomatik Corporation Rat Antithrombin III (AT III) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38686
Encompass_Preferred
Rat Antithrombin Iii (At Iii) Elisa Kit for detecting Antithrombin Iii(At Iii) in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 39-2500ng/ml. Sensitivity: 9.75ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q5M7T5; Serpinc1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8818