Learn More
Biomatik Corporation Rat Apolipoprotein A-IV (APOA4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38695
Encompass_Preferred
Rat Apolipoprotein A-Iv (Apoa4) Elisa Kit for detecting Apolipoprotein A-Iv in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 9.38-600ng/ml. Sensitivity: 2.34ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P02651; Apoa4. Target Synonyms: Apoa4Apolipoprotein A-IV; Apo-AIV; ApoA-IV; Apolipoprotein A4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8820