Learn More
Biomatik Corporation Rat apolipoprotein B-48 (ApoB48) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38696
Encompass_Preferred
Rat Apolipoprotein B-48 (Apob48) Elisa Kit for detecting Apolipoprotein B-48 (Apob48) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ug/ml. Sensitivity: 0.078ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8821