Learn More
Biomatik Corporation Rat brain natriuretic peptide, BNP ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38761
Encompass_Preferred
Rat Brain Natriuretic Peptide, Bnp Elisa Kit for detecting Brain Natriuretic Peptide, Bnp in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 125-8000pg/ml. Sensitivity: 31.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P13205; Nppb. Target Synonyms: NppbNatriuretic peptides B; Gamma-brain natriuretic peptide; Iso-ANP) [Cleaved into: Brain natriuretic peptide 45; 5 kDa cardiac natriuretic peptide; BNP-34)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8833