Learn More
Biomatik Corporation Rat Collagen Type , Col ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38869
Encompass_Preferred
Rat Collagen Type I, Col I Elisa Kit for detecting Collagen Type I, Col I in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: Collagen Type 1; Col 1; Collagen Type I; Col I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8842