Learn More
Biomatik Corporation Rat collagen type (Col ) antibody ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38870
Encompass_Preferred
Rat Collagen Type Ii (Col Ii) Antibody Elisa Kit for detecting Collagen Type II(Col II) Antibody in serum, plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20U/ml. Sensitivity: 0.078u/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: collagen type 2 (Col 2) antibody; collagen type II (Col II) antibody. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8843