Learn More
Biomatik Corporation Rat C-Peptide ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38908
Encompass_Preferred
Rat C-Peptide Elisa Kit for detecting C-Peptide in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.25-20ng/ml. Sensitivity: 1.25ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8846