Learn More
Biomatik Corporation Rat Creatine Kinase MB isoenzyme, CK-MB ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38911
Encompass_Preferred
Rat Creatine Kinase Mb Isoenzyme, Ck-Mb Elisa Kit for detecting Creatine Kinase Mb Isoenzyme, Ck-Mb in serum, urine, tissue homogenates, plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8848