Learn More
Biomatik Corporation Rat CX3C chemokine receptor 1 (CX3CR1) ELISA Kit, 96 tests
Catalog No. 501498851
Encompass_Preferred
Rat Cx3C Chemokine Receptor 1 (Cx3Cr1) Elisa Kit for detecting Chemokine (C-X3-C Motif) Receptor 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P35411; Cx3cr1. Target Synonyms: Cx3cr1; Rbs11; CX3C chemokine receptor 1; C-X3-C CKR-1; CX3CR1; Fractalkine receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8851 Supplier Biomatik Corporation Supplier No. EKC38921
Rat Cx3C Chemokine Receptor 1 (Cx3Cr1) Elisa Kit for detecting Chemokine (C-X3-C Motif) Receptor 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P35411; Cx3cr1. Target Synonyms: Cx3cr1; Rbs11; CX3C chemokine receptor 1; C-X3-C CKR-1; CX3CR1; Fractalkine receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.