Learn More
Biomatik Corporation Rat cytochrome P450 2E1, CYP2E1 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38944
Encompass_Preferred
Rat Cytochrome P450 2E1, Cyp2E1 Elisa Kit for detecting Cytochrome P450, Family 2, Subfamily E, Polypeptide 1 in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P05182; Cyp2e1. Target Synonyms: Cyp2e1; Cyp2e; Cyp2e-1Cytochrome P450 2E1; EC 1.14.14.1; 4-nitrophenol 2-hydroxylase; EC 1.14.13.n7; CYPIIE1; Cytochrome P450-J; Cytochrome P450RLM6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8855