Learn More
Biomatik Corporation Rat dopamine, DA ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38974
Encompass_Preferred
Rat Dopamine, Da Elisa Kit for detecting Dopamine, Da in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8861