Learn More
Biomatik Corporation Rat fatty acid synthase (FAS) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39023
Encompass_Preferred
Rat Fatty Acid Synthase (Fas) Elisa Kit for detecting Fatty Acid Synthase in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P12785; Fasn. Target Synonyms: FasnFatty acid synthase; EC 2.3.1.85) [Includes: [Acyl-carrier-protein] S-acetyltransferase; EC 2.3.1.38); [Acyl-carrier-protein] S-malonyltransferase; EC 2.3.1.39); 3-oxoacyl-[acyl-carrier-protein] synthase; EC 2.3.1.41); 3-oxoacyl-[acyl-carrier-protein] reductase; EC 1.1.1.100); 3-hydroxyacyl-[acyl-carrier-protein] dehydratase; EC 4.2.1.59); Enoyl-[acyl-carrier-protein] reductase; EC 1.3.1.39); Oleoyl-[acyl-carrier-protein] hydrolase; EC 3.1.2.14)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8868