Learn More
Biomatik Corporation rat follicle-stimulating hormone, FSH ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39058
Encompass_Preferred
Rat Follicle-Stimulating Hormone, Fsh Elisa Kit for detecting Follicle-Stimulating Hormone, Fsh in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P18427; Fshb. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8872