Learn More
Biomatik Corporation Rat ghrelin (GHRL) ELISA Kit, 96 tests
Catalog No. 501498876
Encompass_Preferred
Rat Ghrelin (Ghrl) Elisa Kit for detecting Ghrelin/Obestatin Prepropeptide in serum, plasma, cell culture supernates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9QYH7; Ghrl. Target Synonyms: Ghrl; Appetite-regulating hormone; Growth hormone secretagogue; Growth hormone-releasing peptide; Motilin-related peptide) [Cleaved into: Ghrelin; Obestatin-23; Obestatin-13]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8876 Supplier Biomatik Corporation Supplier No. EKC39094
Rat Ghrelin (Ghrl) Elisa Kit for detecting Ghrelin/Obestatin Prepropeptide in serum, plasma, cell culture supernates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9QYH7; Ghrl. Target Synonyms: Ghrl; Appetite-regulating hormone; Growth hormone secretagogue; Growth hormone-releasing peptide; Motilin-related peptide) [Cleaved into: Ghrelin; Obestatin-23; Obestatin-13]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.