Learn More
Biomatik Corporation Rat glutamic acid decarboxylase (GAD) autoantibody (IgG) ELISA Kit, 96 tests
Catalog No. 501498882
Encompass_Preferred
Rat Glutamic Acid Decarboxylase (Gad) Autoantibody (Igg) Elisa Kit for detecting Glutamic Acid Decarboxylase Autoantibody Igg, Gad-Ab-Igg in serum, plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8882 Supplier Biomatik Corporation Supplier No. EKC39113
Rat Glutamic Acid Decarboxylase (Gad) Autoantibody (Igg) Elisa Kit for detecting Glutamic Acid Decarboxylase Autoantibody Igg, Gad-Ab-Igg in serum, plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.