Learn More
Biomatik Corporation Rat guanylate cyclase activator 2B (uroguanylin) (GUCA2B) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39157
Encompass_Preferred
Rat Guanylate Cyclase Activator 2B (Uroguanylin) (Guca2B) Elisa Kit for detecting Guanylate Cyclase Activator 2B (Uroguanylin) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P70668; Guca2b. Target Synonyms: Guca2b; Guanylate cyclase activator 2B [Cleaved into: Uroguanylin; UGN)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8887